2-(1H-Pyrrol-1-yl)-1-ethanamine 95%

Supplier: Ambeed
Warning

Synonyms: 2-(1H-Pyrrol-1-yl)ethanamine

A215921-100MG A215921-1G A215921-250MG A215921-10G
77237-552EA 29.87 USD
77237-552 77237-554 77237-556 77746-299
2-(1H-Pyrrol-1-yl)-1-ethanamine 95%
2-(1H-Pyrrol-1-yl)-1-ethanamine

Precaución: for Research Use Only

Formula: C₆H₁₀N₂
MW: 110.16 g/mol
MDL Number: MFCD00958089
CAS Number: 29709-35-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR