3-Bromo-5-methylpyrazin-2-amine 95%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-3-bromo-5-methylpyrazine

A215877-100MG A215877-1G A215877-250MG
77249-220EA 21.63 USD
77249-220 77249-222 77249-224
3-Bromo-5-methylpyrazin-2-amine 95%
3-Bromo-5-methylpyrazin-2-amine

Precaución: for Research Use Only

Formula: C5H6BrN3
MW: 188.0300 g/mol
MDL Number: MFCD09750037
CAS Number: 74290-65-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR