1,3,5-Tris(4-biphenylyl)benzene 98+%

Supplier: Ambeed
Warning

Synonyms: 5''-([1,1'-Biphenyl]-4-yl)-1,1':4',1'':3'',1''':4''',1''''-quinquephenyl

A215810-1G A215810-250MG
77237-548EA 67 USD
77237-548 77237-550
1,3,5-Tris(4-biphenylyl)benzene 98+%
1,3,5-Tris(4-biphenylyl)benzene

Precaución: for Research Use Only

Formula: C₄₂H₃₀
MW: 534.70 g/mol
MDL Number: MFCD23382303
CAS Number: 6326-64-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR