4-Methylphenylacetone 98%

Supplier: Ambeed
Warning

Synonyms: 1-(p-Tolyl)propan-2-one

A215590-1G A215590-250MG
77288-092EA 58.71 USD
77288-092 77288-094
4-Methylphenylacetone 98%
4-Methylphenylacetone

Precaución: for Research Use Only

Formula: C₁₀H₁₂O
MW: 148.20 g/mol
MDL Number: MFCD00017250
CAS Number: 2096-86-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR