4-(Bromomethyl)benzaldehyde 97%

Supplier: Ambeed
Danger

A215549-1G A215549-250MG A215549-5G A215549-25G
77288-084EA 40.17 USD
77288-084 77288-086 77550-392 77568-080
4-(Bromomethyl)benzaldehyde 97%
4-(Bromomethyl)benzaldehyde

Precaución: For research use only.

Formula: C₈H₇BrO
MDL Number: MFCD08271963
CAS Number: 51359-78-5
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR