1,1,1,3,3,3-Hexafluoropropan-2-yl 4-(bis(benzo[d][1,3]dioxol-5-yl)(hydroxy)methyl)piperidine-1-carboxylate ≥98%

Supplier: Ambeed
Warning

A215224-10MG A215224-5MG
77237-534EA 121 USD
77237-534 77237-536 77237-538
1,1,1,3,3,3-Hexafluoropropan-2-yl 4-(bis(benzo[d][1,3]dioxol-5-yl)(hydroxy)methyl)piperidine-1-carboxylate ≥98%
1,1,1,3,3,3-Hexafluoropropan-2-yl 4-(bis(benzo[d][1,3]dioxol-5-yl)(hydroxy)methyl)piperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₂₄H₂₁F₆NO₇
MDL Number: MFCD22987957
CAS Number: 1380424-42-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR