3-Fluorobenzoyl chloride 98%

Supplier: Ambeed
Danger

A215170-25G A215170-5G
77275-282EA 23 USD
77275-282 77275-284
3-Fluorobenzoyl chloride 98%
3-Fluorobenzoyl chloride

Precaución: for Research Use Only

Formula: C₇H₄ClFO
MDL Number: MFCD00000670
CAS Number: 1711-07-5
UN: 3265
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR