4-Methoxy-1-butanol 98%

Supplier: Ambeed
Danger

Synonyms: 4-Methoxybutan-1-ol

A214893-1G A214893-250MG A214893-25G A214893-5G
77288-074EA 9.27 USD
77288-074 77288-076 77288-078 77288-080
4-Methoxy-1-butanol 98%
4-Methoxy-1-butanol

Precaución: for Research Use Only

Formula: C₅H₁₂O₂
MW: 104.15 g/mol
MDL Number: MFCD00040437
CAS Number: 111-32-0
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR