3,5-Dibenzyloxybenzyl alcohol 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
(3,5-Bis(benzyloxy)phenyl)methanol, 3,5-Bis(benzyloxy)benzyl alcohol
A214615-1G
A214615-25G
A214615-5G
77237-518EA
59
USD
77237-518
77237-520
77237-522
3,5-Dibenzyloxybenzyl alcohol 97%
3,5-Dibenzyloxybenzyl alcohol
Precaución: for Research Use Only
Formula:
C₂₁H₂₀O₃ |
MDL Number:
MFCD01863236 CAS Number: 24131-31-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...