2-Bromo-9,9-diethylfluorene 98%

Supplier: Ambeed
Warning

Synonyms: 2-Bromo-9,9-diethyl-9H-fluorene

A214487-1G A214487-250MG
77261-172EA 21.63 USD
77261-172 77261-174
2-Bromo-9,9-diethylfluorene 98%
2-Bromo-9,9-diethylfluorene

Precaución: for Research Use Only

Formula: C₁₇H₁₇Br
MW: 301.23 g/mol
MDL Number: MFCD09909863
CAS Number: 287493-15-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR