1-Bromo-2-butyne 97%

Supplier: Ambeed
Danger

A214308-10G A214308-1G A214308-25G A214308-5G
77237-506EA 14.42 USD
77237-506 77237-508 77237-510 77237-512
1-Bromo-2-butyne 97%
1-Bromo-2-butyne

Precaución: for Research Use Only

Formula: C₄H₅Br
MW: 132.99 g/mol
MDL Number: MFCD00190233
CAS Number: 3355-28-0
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR