2-(2-Methyl-1H-imidazol-1-yl)benzoic acid 97%

Supplier: Ambeed
Warning

Synonyms: 1-(2-Carboxyphenyl)-2-methyl-1H-imidazole

A214084-100MG A214084-250MG
77237-474EA 37.08 USD
77237-474 77237-476
2-(2-Methyl-1H-imidazol-1-yl)benzoic acid 97%
2-(2-Methyl-1H-imidazol-1-yl)benzoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₀N₂O₂
MW: 202.209 g/mol
MDL Number: MFCD09065012
CAS Number: 159589-71-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR