tert-Butyl 2-(3-bromophenyl)acetate 95%

Supplier: Ambeed
Warning

A213991-100MG A213991-1G A213991-250MG A213991-5G
77237-466EA 24.72 USD
77237-466 77237-468 77237-470 77237-472
tert-Butyl 2-(3-bromophenyl)acetate 95%
tert-Butyl 2-(3-bromophenyl)acetate

Precaución: for Research Use Only

Formula: C₁₂H₁₅BrO₂
MW: 271.15 g/mol
MDL Number: MFCD19026476
CAS Number: 197792-52-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR