Cytidine-5'-monophosphate 95%
Supplier: Ambeed
|
Warning
|
A213941-100G
A213941-10G
A213941-25G
A213941-500G
A213941-5G
77249-184EA
37.08
USD
77249-184
77249-186
77249-188
77249-190
77249-192
Cytidine-5'-monophosphate 95%
Cytidine-5'-monophosphate
Precaución: for Research Use Only
Formula:
C₉H₁₄N₃O₈P MW: 323.20 g/mol |
MDL Number:
MFCD00006544 CAS Number: 63-37-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...