3,5-Difluorobenzonitrile 98%

Supplier: Ambeed
Warning

A213922-100G A213922-10G A213922-25G A213922-500G A213922-5G
77275-252EA 59.74 USD
77275-252 77275-254 77275-256 77275-258 77275-260
3,5-Difluorobenzonitrile 98%
3,5-Difluorobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₃F₂N
MW: 139.1 g/mol
MDL Number: MFCD00010311
CAS Number: 64248-63-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR