4,4,5,5,5-Pentafluoropentan-1-ol 98%

Supplier: Ambeed
Warning

A213899-1G A213899-25G A213899-5G
77288-048EA 8.24 USD
77288-048 77288-050 77288-052
4,4,5,5,5-Pentafluoropentan-1-ol 98%
4,4,5,5,5-Pentafluoropentan-1-ol

Precaución: for Research Use Only

Formula: C₅H₇F₅O
MW: 178.1 g/mol
MDL Number: MFCD00153224
CAS Number: 148043-73-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR