3'-Amino-4'-fluoroacetophenone 98%

Supplier: Ambeed
Warning

A213810-10G A213810-1G A213810-250MG A213810-5G
77288-042EA 410.97 USD
77288-042 77288-044 77288-046 77748-334
3'-Amino-4'-fluoroacetophenone 98%
3'-Amino-4'-fluoroacetophenone

Precaución: for Research Use Only

Formula: C₈H₈FNO
MW: 153.16 g/mol
MDL Number: MFCD00115400
CAS Number: 2002-82-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR