2,2-Dimethyl-3-(m-tolyl)propan-1-ol 98%

Supplier: Ambeed
Warning

A213633-100G A213633-10G A213633-25G A213633-500G
77249-176EA 27.81 USD
77249-176 77249-178 77249-180 77249-182
2,2-Dimethyl-3-(m-tolyl)propan-1-ol 98%
2,2-Dimethyl-3-(m-tolyl)propan-1-ol

Precaución: for Research Use Only

Formula: C₁₂H₁₈O
MW: 178.27 g/mol
MDL Number: MFCD03701092
CAS Number: 103694-68-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR