2-(2-Chloroethoxy)ethanol 97%

Supplier: Ambeed
Warning

A213420-100G A213420-500G A213420-25G
77237-440EA 147.29 USD
77237-440 77237-442 77237-444 77303-288
2-(2-Chloroethoxy)ethanol 97%
2-(2-Chloroethoxy)ethanol

Precaución: for Research Use Only

Formula: C₄H₉ClO₂
MDL Number: MFCD00002870
CAS Number: 628-89-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR