ACY-738 98%

Supplier: Ambeed
Warning

Synonyms: N-Hydroxy-2-((1-phenylcyclopropyl)amino)pyrimidine-5-carboxamide

A213388-1MG A213388-2MG A213388-10MG A213388-50MG A213388-5MG A213388-100MG A213388-250MG
77237-436EA 10.3 USD
77237-436 77237-438 77615-804 77615-806 77615-808 77746-290 77746-291
ACY-738 98%
ACY-738

Precaución: for Research Use Only

Formula: C₁₄H₁₄N₄O₂
MW: 270.29 g/mol
MDL Number: MFCD28347706
CAS Number: 1375465-91-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR