GW 4869 dihydrocloride 97%

Supplier: Ambeed
Warning

Synonyms: GW4869, p-Benzenediacrylanilide, 4′,4′′-di-2-imidazolin-2-yl-, dihydrochloride

A213331-10MG A213331-1MG A213331-25MG A213331-2MG A213331-50MG A213331-5MG
77261-120EA 146 USD
77261-120 77261-122 77261-124 77261-126 77261-128 77261-130
GW 4869 dihydrocloride 97%
GW 4869 dihydrocloride

Precaución: for Research Use Only

Formula: C₃₀H₃₀Cl₂N₆O₂
MDL Number: MFCD06411564
CAS Number: 6823-69-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR