3-Methyl-2-nitrobenzyl alcohol 98%

Supplier: Ambeed
Warning

Synonyms: (3-Methyl-2-nitrophenyl)methanol, 3-(Hydroxymethyl)-2-nitrotoluene

A213318-1G A213318-250MG A213318-5G
77249-170EA 32.96 USD
77249-170 77249-172 77249-174
3-Methyl-2-nitrobenzyl alcohol 98%
3-Methyl-2-nitrobenzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₉NO₃
MW: 167.16 g/mol
MDL Number: MFCD00007182
CAS Number: 80866-76-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR