5,5'-(Pyridine-2,5-diyl)diisophthalic acid 98%

Supplier: Ambeed
Warning

A213104-100MG A213104-1G A213104-250MG
77261-110EA 10 USD
77261-110 77261-112 77261-114
5,5'-(Pyridine-2,5-diyl)diisophthalic acid 98%
5,5'-(Pyridine-2,5-diyl)diisophthalic acid

Precaución: for Research Use Only

Formula: C₂₁H₁₃NO₈
CAS Number: 1431292-15-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR