CHR2797 98%

Supplier: Ambeed
Warning

Synonyms: alpha-[[(2R)-2-[(1S)-1-Hydroxy-2-(hydroxyamino)-2-oxoethyl]-4-methyl-1-oxopentyl]amino]benzeneacetic acid cyclopentyl ester, Cyclopentyl (S)-2-((R)-2-((S)-1-hydroxy-2-(hydroxyamino)-2-oxoethyl)-4-methylpentanamido)-2-phenylacetate, Tosedostat

A212860-1MG A212860-5MG A212860-10MG
77249-150EA 42 USD
77249-150 77615-802 77615-800
CHR2797 98%
CHR2797

Precaución: for Research Use Only

Formula: C₂₁H₃₀N₂O₆
MW: 406.5 g/mol
MDL Number: MFCD13185162
CAS Number: 238750-77-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR