1H,1H-Pentadecafluoro-1-octanol 98%

Supplier: Ambeed
Warning

Synonyms: 1H,1H-Perfluoro-1-octanol, 1H,1h-pentadecafluoro-1-octanol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-Pentadecafluor-1-octanol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-Pentadecafluoro-1-octanol

A212704-1G A212704-250MG A212704-5G
77237-424EA 18.54 USD
77237-424 77237-426 77237-428
1H,1H-Pentadecafluoro-1-octanol 98%
1H,1H-Pentadecafluoro-1-octanol

Precaución: for Research Use Only

Formula: C₈H₃F₁₅O
MW: 400.09 g/mol
MDL Number: MFCD00004675
CAS Number: 307-30-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR