4-Fluoro-4'-hydroxybenzophenone 98%

Supplier: Ambeed
Warning

Synonyms: (4-Fluorophenyl)(4-hydroxyphenyl)methanone

A212527-100G A212527-1G A212527-25G A212527-5G
77287-994EA 406.01 USD
77287-994 77287-996 77287-998 77288-000
4-Fluoro-4'-hydroxybenzophenone 98%
4-Fluoro-4'-hydroxybenzophenone

Precaución: for Research Use Only

Formula: C₁₃H₉FO₂
MDL Number: MFCD00011484
CAS Number: 25913-05-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR