6-Chloro-2-oxoindoline-5-carboxylic acid 97%

Supplier: Ambeed
Warning

A212508-100MG A212508-250MG A212508-1G
77261-092EA 64.89 USD
77261-092 77261-094 77748-323
6-Chloro-2-oxoindoline-5-carboxylic acid 97%
6-Chloro-2-oxoindoline-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₆ClNO₃
MW: 211.60 g/mol
MDL Number: MFCD12174764
CAS Number: 1156390-49-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR