(2-Dicyclohexylphosphino-2',6'-dimethoxybiphenyl)[2-(2'-amino-1,1'-biphenyl)]palladium(II) methanesulfonate 97%

Supplier: Ambeed
Warning

A212507-100MG A212507-1G A212507-250MG A212507-5G A212507-25G
77249-136EA 7 USD
77249-136 77249-138 77249-140 77249-142 77746-288
(2-Dicyclohexylphosphino-2',6'-dimethoxybiphenyl)[2-(2'-amino-1,1'-biphenyl)]palladium(II) methanesulfonate 97%
(2-Dicyclohexylphosphino-2',6'-dimethoxybiphenyl)[2-(2'-amino-1,1'-biphenyl)]palladium(II) methanesulfonate

Precaución: for Research Use Only

Formula: C₄₀H₅₁NO₈PPdS₂
MDL Number: MFCD29472404
CAS Number: 1445085-82-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR