(S)-2-Amino-2-(naphthalen-1-yl)acetic acid hydrochloride 97%

Supplier: Ambeed
Warning

A212387-100MG A212387-1G
77261-088EA 58 USD
77261-088 77261-090
(S)-2-Amino-2-(naphthalen-1-yl)acetic acid hydrochloride 97%
(S)-2-Amino-2-(naphthalen-1-yl)acetic acid hydrochloride

Precaución: for Research Use Only

Formula: C₁₂H₁₂ClNO₂
MDL Number: MFCD22201196
CAS Number: 649554-52-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR