Methyl 2-amino-5-chloronicotinate 95%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-amino-5-chloropyridine-3-carboxylate, 2-Amino-5-chloro-3-(methoxycarbonyl)pyridine

A212277-100MG A212277-10G A212277-1G A212277-250MG A212277-5G
77249-126EA 5.15 USD
77249-126 77249-128 77249-130 77249-132 77249-134
Methyl 2-amino-5-chloronicotinate 95%
Methyl 2-amino-5-chloronicotinate

Precaución: for Research Use Only

Formula: C₇H₇ClN₂O₂
MW: 186.596 g/mol
MDL Number: MFCD09031678
CAS Number: 50735-33-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR