(5-Bromo-2-methylpyridin-3-yl)methanol 98%

Supplier: Ambeed
Warning

A212036-100MG A212036-250MG
77237-404EA 67.98 USD
77237-404 77237-406
(5-Bromo-2-methylpyridin-3-yl)methanol 98%
(5-Bromo-2-methylpyridin-3-yl)methanol

Precaución: for Research Use Only

Formula: C₇H₈BrNO
MW: 202.04 g/mol
MDL Number: MFCD26518812
CAS Number: 1346541-53-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR