3-Aminopyrazine-2-carboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3-Amino-2-pyrazinecarboxylic acid

A211851-100G A211851-25G A211851-500G A211851-5G A211851-10G
77275-194EA 65 USD
77275-194 77275-196 77275-198 77275-200 77748-320
3-Aminopyrazine-2-carboxylic acid 98%
3-Aminopyrazine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₅H₅N₃O₂
MDL Number: MFCD00006141
CAS Number: 5424-01-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR