4-Ethyl-N-(7-methoxy-4-((3-vinylphenyl)amino)quinazolin-6-yl)piperazine-1-carboxamide 97%

Supplier: Ambeed
Warning

A211761-100MG A211761-1G A211761-250MG
77261-080EA 63 USD
77261-080 77261-082 77261-084
4-Ethyl-N-(7-methoxy-4-((3-vinylphenyl)amino)quinazolin-6-yl)piperazine-1-carboxamide 97%
4-Ethyl-N-(7-methoxy-4-((3-vinylphenyl)amino)quinazolin-6-yl)piperazine-1-carboxamide

Precaución: for Research Use Only

Formula: C₂₄H₂₈N₆O₂
CAS Number: 2109805-51-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR