2-Nitro-5-(phenylthio)aniline 97%

Supplier: Ambeed
Warning

A211718-100G A211718-10G A211718-1G A211718-250MG A211718-25G A211718-5G
77275-182EA 338.87 USD
77275-182 77275-184 77275-186 77275-188 77275-190 77275-192
2-Nitro-5-(phenylthio)aniline 97%
2-Nitro-5-(phenylthio)aniline

Precaución: for Research Use Only

Formula: C₁₂H₁₀N₂O₂S
MW: 246.28 g/mol
MDL Number: MFCD00129835
CAS Number: 43156-47-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR