4,4'-Bis(2,2-diphenylvinyl)biphenyl 99%

Supplier: Ambeed
Warning

Synonyms: 4,4'-Bis(2,2-diphenylvinyl)-1,1'-biphenyl, DPVBi

A211592-100MG A211592-1G
77287-976EA 17.51 USD
77287-976 77287-978
4,4'-Bis(2,2-diphenylvinyl)biphenyl 99%
4,4'-Bis(2,2-diphenylvinyl)biphenyl

Precaución: for Research Use Only

Formula: C₄₀H₃₀
MW: 510.68 g/mol
MDL Number: MFCD03093280
CAS Number: 142289-08-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR