2-Methoxy-5-(trifluoromethyl)pyridine 97%

Supplier: Ambeed
Warning

A211546-1G A211546-5G A211546-10G
77275-168EA 9 USD
77275-168 77275-170 77748-316
2-Methoxy-5-(trifluoromethyl)pyridine 97%
2-Methoxy-5-(trifluoromethyl)pyridine

Precaución: for Research Use Only

Formula: C₇H₆F₃NO
MDL Number: MFCD00153203
CAS Number: 175277-45-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR