2-Amino-3H-pyrrolo[3,2-d]pyrimidin-4(5H)-one 97%

Supplier: Ambeed
Warning

A211545-100MG A211545-1G A211545-250MG
77287-970EA 58.71 USD
77287-970 77287-972 77287-974
2-Amino-3H-pyrrolo[3,2-d]pyrimidin-4(5H)-one 97%
2-Amino-3H-pyrrolo[3,2-d]pyrimidin-4(5H)-one

Precaución: for Research Use Only

Formula: C₆H₆N₄O
MW: 150.14 g/mol
MDL Number: MFCD09540493
CAS Number: 65996-58-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR