2-(4-Bromophenyl)imidazo[1,2-a]pyridine 97%

Supplier: Ambeed
Warning

A211423-10G A211423-1G A211423-250MG A211423-5G
77237-390EA 201 USD
77237-390 77237-392 77237-394 77237-396
2-(4-Bromophenyl)imidazo[1,2-a]pyridine 97%
2-(4-Bromophenyl)imidazo[1,2-a]pyridine

Precaución: for Research Use Only

Formula: C₁₃H₉BrN₂
MDL Number: MFCD00218250
CAS Number: 34658-66-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR