(2-((Dimethylamino)methyl)phenyl)boronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-(N,N-Dimethylaminomethyl)phenylboronic acid, 2-(Dimethylaminomethyl)phenylboronic acid, 2-(Dimethylaminomethyl)benzeneboronic acid

A211289-10G A211289-1G A211289-250MG A211289-25G A211289-5G
77249-104EA 6.18 USD
77249-104 77249-106 77249-108 77249-110 77249-112 77249-114
(2-((Dimethylamino)methyl)phenyl)boronic acid 98%
(2-((Dimethylamino)methyl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₉H₁₄BNO₂
MDL Number: MFCD01318999
CAS Number: 85107-53-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR