(Pyrrolidin-3-yl)acetic acid hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: 3-(Carboxymethyl)pyrrolidine hydrochloride, 3-(Carboxymethyl)tetrahydropyrrole hydrochloride, 2-(Pyrrolidin-3-yl)acetic acid hydrochloride

A211203-100MG A211203-1G A211203-5G
77287-964EA 37.08 USD
77287-964 77287-966 77287-968
(Pyrrolidin-3-yl)acetic acid hydrochloride 98%
(Pyrrolidin-3-yl)acetic acid hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₁NO₂·ClH
MW: 165.618 g/mol
MDL Number: MFCD06795694
CAS Number: 80616-50-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR