4-(Methylthio)phenol 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-(Methylmercapto)phenol
A211198-100G
A211198-10G
A211198-1G
A211198-25G
A211198-5G
77275-156EA
99.91
USD
77275-156
77275-158
77275-160
77275-162
77275-164
4-(Methylthio)phenol 98%
4-(Methylthio)phenol
Precaución: for Research Use Only
Formula:
C₇H₈OS MW: 140.21 g/mol |
MDL Number:
MFCD00002351 CAS Number: 1073-72-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...