2,3,5,6-Tetrafluorobenzonitrile 92%

Supplier: Ambeed
Danger

A210906-1G A210906-250MG A210906-5G
77249-094EA 11 USD
77249-094 77249-096 77249-098
2,3,5,6-Tetrafluorobenzonitrile 92%
2,3,5,6-Tetrafluorobenzonitrile

Precaución: for Research Use Only

Formula: C₇HF₄N
MDL Number: MFCD00012286
CAS Number: 5216-17-1
UN: 3439
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR