7-Oxo-7H-benzo[de]benzo[4,5]imidazo[2,1-a]isoquinoline-3-carboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: 7-Oxo-7H-benzo[de]benzo[4,5]imidazo[2,1-a]isoquinoline-3-carboxylic acid

A210855-10MG A210855-1MG A210855-25MG A210855-50MG A210855-5MG
77261-044EA 46.35 USD
77261-044 77261-046 77261-048 77261-050 77261-052
7-Oxo-7H-benzo[de]benzo[4,5]imidazo[2,1-a]isoquinoline-3-carboxylic acid 98%
STO-609

Precaución: for Research Use Only

Formula: C₁₉H₁₀N₂O₃
MW: 314.29 g/mol
MDL Number: MFCD30685721
CAS Number: 52029-86-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR