5-Phenyl-5,12-dihydroindolo[3,2-a]carbazole 98%

Supplier: Ambeed
Warning

A210775-1G A210775-250MG A210775-25G
77249-088EA 25.75 USD
77249-088 77249-090 77249-092
5-Phenyl-5,12-dihydroindolo[3,2-a]carbazole 98%
5-Phenyl-5,12-dihydroindolo[3,2-a]carbazole

Precaución: for Research Use Only

Formula: C₂₄H₁₆N₂
MW: 332.41 g/mol
MDL Number: MFCD28975052
CAS Number: 1247053-55-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR