1-(1H-Indol-2-yl)ethanone 98%

Supplier: Ambeed
Warning

Synonyms: 2-Acetylindole

A210759-1G A210759-250MG A210759-5G
77249-082EA 74.16 USD
77249-082 77249-084 77249-086
1-(1H-Indol-2-yl)ethanone 98%
1-(1H-Indol-2-yl)ethanone

Precaución: for Research Use Only

Formula: C₁₀H₉NO
MW: 159.19 g/mol
MDL Number: MFCD01863329
CAS Number: 4264-35-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR