Belinostat 98%

Supplier: Ambeed
Warning

A210633-100MG A210633-1MG A210633-1G
77237-382EA 36 USD
77237-382 77237-384 77746-282
Belinostat 98%
Belinostat

Precaución: for Research Use Only

Formula: C₁₅H₁₄N₂O₄S
MDL Number: MFCD08064035
CAS Number: 866323-14-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR