2,3-Dichloro-5,8-dihydroxy-1,4-naphthoquinone 97%

Supplier: Ambeed
Warning

Synonyms: 2,3-Dichloro-5,8-dihydroxynaphthalene-1,4-dione

A210622-100MG A210622-250MG
77249-078EA 27.81 USD
77249-078 77249-080
2,3-Dichloro-5,8-dihydroxy-1,4-naphthoquinone 97%
2,3-Dichloro-5,8-dihydroxy-1,4-naphthoquinone

Precaución: for Research Use Only

Formula: C₁₀H₄Cl₂O₄
MW: 259.04 g/mol
MDL Number: MFCD00075261
CAS Number: 14918-69-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR