Amifostine trihydrate 97%

Supplier: Ambeed
Warning

Synonyms: S-(2-((3-Aminopropyl)amino)ethyl) O,O-dihydrogen phosphorothioate trihydrate

A210496-100MG A210496-10MG A210496-1MG A210496-250MG A210496-50MG A210496-5G A210496-1G
77261-024EA 40 USD
77261-024 77261-026 77261-028 77261-030 77261-032 77261-034 77615-778
Amifostine trihydrate 97%
Amifostine trihydrate

Precaución: for Research Use Only

Formula: C₅H₁₅N₂O₃PS·₃H₂O
MW: 268.27 g/mol
MDL Number: MFCD00453866
CAS Number: 112901-68-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR