4-Cyclopropyl-1H-pyrazol-3-amine 95%

Supplier: Ambeed
Warning

A210486-100MG A210486-1G A210486-250MG
77275-142EA 98.88 USD
77275-142 77275-144 77275-146
4-Cyclopropyl-1H-pyrazol-3-amine 95%
4-Cyclopropyl-1H-pyrazol-3-amine

Precaución: for Research Use Only

Formula: C₆H₉N₃
MW: 123.156 g/mol
MDL Number: MFCD08752485
CAS Number: 673475-74-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR