6-Fluorochromane-2-carboxylic acid 97%

Supplier: Ambeed
Warning

A210260-1G A210260-250MG A210260-25G A210260-5G
77261-012EA 10 USD
77261-012 77261-014 77261-016 77261-018
6-Fluorochromane-2-carboxylic acid 97%
6-Fluorochromane-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₉FO₃
MDL Number: MFCD07778357
CAS Number: 99199-60-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR